Entrectinib 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
474611000
474611000.EA
229
EUR
474611000.
Entrectinib 98%
Entrectinib
Formel:
C₃₁H₃₄F₂N₆O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28129099 CAS-Nummer: 1108743-60-7 |
Spezifikation Testergebnisse
Appearance | white to light beige powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...