Erdafitinib 98+%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N1-(3,5-dimethoxyphenyl)-N2-isopropyl-N1-(3-(1-methyl-1H-pyrazol-4-yl)quinoxalin-6-yl)ethane-1,2-diamine, JNJ-42756493
474621000
474621000.EA
229
EUR
474621000.
Erdafitinib 98+%
Erdafitinib
Formel:
C₂₅H₃₀N₆O₂ Molecular Weight: 446,55 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12352005 CAS-Nummer: 1346242-81-6 |
Spezifikation Testergebnisse
Appearance | white to yellow powder |
Infrared spectrum | Conforms |
HPLC | ≥98.0 % |
Melting point | 136°C to 141°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...