Fedratinib (TG101348) 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-(tert-Butyl)-3-((5-methyl-2-((4-(2-(pyrrolidin-1-yl)ethoxy)phenyl)amino)pyrimidin-4-yl)amino)benzenesulphonamide, Fedratinib, SAR-302503

474641000
474641000.EA 257 EUR
474641000.
Fedratinib (TG101348) 98%
Fedratinib (TG101348)
Formel: C₂₇H₃₆N₆O₃S
Molecular Weight: 524,7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12922515
CAS-Nummer: 936091-26-8

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white solid
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR