Gilteritinib 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Ethyl-3-((3-methoxy-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)phenyl)amino)-5-((tetrahydro-2H-pyran-4-yl)amino)pyrazine-2-carboxamide, ASP 2215

474651000
474651000.EA 229 EUR
474651000.
Gilteritinib 98%
Gilteritinib
Formel: C₂₉H₄₄N₈O₃
Molecular Weight: 552,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28144685
CAS-Nummer: 1254053-43-4

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow solid
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR