Momelotinib 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-(Cyanomethyl)-4-(2-((4-morpholinophenyl)amino)pyrimidin-4-yl)benzamide, CYT387

474681000
474681000.EA 141 EUR
474681000.
Momelotinib 97%
Momelotinib
Formel: C₂₃H₂₂N₆O₂
Molecular Weight: 414,5 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16038899
CAS-Nummer: 1056634-68-4

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow solid
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR