Selpercatinib 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-(2-Hydroxy-2-methylpropoxy)-4-(6-(6-((6-methoxypyridin-3-yl)methyl)-3,6-diazabicyclo[3.1.1]heptan-3-yl)pyridin-3-yl)pyrazolo[1,5-a]pyridine-3-carbonitrile

474731000
474731000.EA 115 EUR
474731000.
Selpercatinib 98%
Selpercatinib
Formel: C₂₉H₃₁N₇O₃
Molecular Weight: 525,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31814089
CAS-Nummer: 2152628-33-4

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white solid
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR