EMD-1214063 98%

Lieferant: Thermo Scientific

Synonyme: Tepotinib, 3-(1-(3-(5-((1-Methylpiperidin-4-yl)methoxy)pyrimidin-2-yl)benzyl)-6-oxo-1,6-dihydropyridazin-3-yl)benzonitrile, 3-[1-[[3-[5-[(1-methylpiperidin-4-yl)methoxy]pyrimidin-2-yl]phenyl]methyl]-6-oxopyridazin-3-yl]benzonitrile

474741000
474741000.EA 514 EUR
474741000.
EMD-1214063 98%
EMD-1214063
Formel: C₂₉H₂₈N₆O₂
Molecular Weight: 492,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18452823
CAS-Nummer: 1100598-32-0

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white solid
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR