Colecalciferol (Vitamin D3)

Lieferant: Merck

Synonyme: Calciol, (3β,5Z,7E)-9,10-Secocholesta- 5,7,10(19)-trien-3-ol, Vitamin D3, 3β,5-cis,7-trans-3-Hydroxy-9,10-secocholesta- 5,7,10(19)-trien, Colecalciferol, 3-[2-[7a-Methyl-1-(6-methylheptan-2-yl)- 2,3,3a,5,6,7-hexahydro-1H-inden-4-yliden] ethyliden]-4-methyliden-cyclohexan-1-ol

47763
47763.EA 60.2 EUR
47763.
Colecalciferol (Vitamin D3)
Colecalciferol (Vitamin D3)
Formel: C₂₇H₄₄O
Molecular Weight: 384,65 g/mol
MDL: MFCD00078131
CAS-Nummer: 67-97-0
EINECS: 200-673-2
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=976bbc19d9053110b55cc25575a6ca52edc9e649&vid=976bbc19d9053110b55cc25575a6ca52edc9e649&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR