DL-α-Tocopherol

Lieferant: Merck

47783
47783.EA 60.2 EUR
47783.
DL-α-Tocopherol
DL-α-Tocopherol
Formel: C₂₉H₅₀O₂
MDL: MFCD00072051
CAS-Nummer: 10191-41-0
EINECS: 233-466-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da15a597e3aa562597de5daeca96176dcd44e2a1&vid=da15a597e3aa562597de5daeca96176dcd44e2a1&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR