Riboflavin (Vitamin B2)

Lieferant: Merck

47861
47861.EA 43 EUR
47861.
Riboflavin (Vitamin B2)
Riboflavin (Vitamin B2)
Formel: C₁₇H₂₀N₄O₆
Schmelzpunkt: 280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005022
CAS-Nummer: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eabefeb1edc9c07a64c5aa2c619b34b6fc0c8b3c&vid=eabefeb1edc9c07a64c5aa2c619b34b6fc0c8b3c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR