Pyridoxin Hydrochlorid

Lieferant: Merck

Synonyme: Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI

47862
47862.EA 43 EUR
47862.
Pyridoxin Hydrochlorid
Pyridoxin Hydrochlorid
Formel: C₈H₁₂ClNO₃
Molecular Weight: 205,64 g/mol
Schmelzpunkt: 214…215 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012807
CAS-Nummer: 58-56-0
EINECS: 200-386-2
Merck Index: 13,08072
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=804baf242a846828039e8d36eb3055ad88698d28&vid=804baf242a846828039e8d36eb3055ad88698d28&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR