Vitamin B12 (Cyanocobalamin)

Lieferant: Merck

47869
47869.EA 54.1 EUR
47869.
Vitamin B12 (Cyanocobalamin)
Vitamin B12 (Cyanocobalamin)
Formel: C₆₃H₈₈CoN₁₄O₁₄P
Molecular Weight: 1355,38 g/mol
Schmelzpunkt: 320 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00151092
CAS-Nummer: 68-19-9
EINECS: 200-680-0
Merck Index: 12,10152
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78d2c09d5cca0f19fff2fb63ddf68d8ad9fc1c0e&vid=78d2c09d5cca0f19fff2fb63ddf68d8ad9fc1c0e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR