Barcode-Lesegerät
Lieferant: THERMO MATRIX TECHNOLOGIES
3125
479-1013EA 2421 EUR
479-1013
Barcode-Lesegerät
Barcode- und RFID-Systeme
The VisionMate ST 2D barcode reader is a compact, single tube reader for scanning 2D barcoded tubes into virtually any application.
  • Directly import your scanned barcode data into any Microsoft® Windows® application
  • Export via Comma Separated Variable file, Microsoft Excel®, XML, COM port, ODBC or TCP socket
  • Simple to install and use
  • For individual sample retrieval
Simply plug the barcode reader into an available USB port and allow the drive to automatically install. Hold any Thermo Scientific 2D barcoded storage tube over the scanning window to decode the barcode instantly.