Kryoröhrchen
Lieferant: Brand
114830 114831 114832 114833 114834 114835 114840 114841 114842 114843 114844 114845
479-1201EA 336 EUR
479-1201 479-1202 479-1203 479-1204 479-1205 479-1206 479-1211 479-1212 479-1213 479-1214 479-1215 479-1216
Kryoröhrchen
Vials
PP, graduated. Designed for storage of biological material, such as microorganisms, human and animal cells, more. in the gaseous phase of liquid nitrogen.
  • Large frosted marking area and coloured cap inserts for easy sample identification
  • Temperature stability to −196 °C and autoclavable at 121 °C (2 bar), acc. DIN EN 285
  • γ-ray sterile (SAL 10⁻⁶)
  • Cryogenic tubes are sterile, RNase-, DNase-, DNA- and endotoxin-free
  • Tubes without ring stands can be centrifuged at up to an RCF of 14000×g
Freistehend
* mit rundem Boden