Horizontal and vertical autoclave with automatic control. The chamber is constructed of long lasting 316L/Ti grade stainless steel with superior corrosion resistance; the generator is made from stainless steel.
- Micro processor control with fully automatic cycles and process control
- Easy operating
- Flexible temperature probe to measure in the load
- Six (6) variable programs (14 variable programs for (F)ull (O)ption ELV- Models)
- Short cycle times due to intelligent pressure- and temperature control
- Pressure- and temperature controlled door safety locking
- Fully automatic feed water control (automatic water filling for the chamber by magnetic valve)
- Built-in steam generator safety: A water level monitoring system maintains a constant water level and ensures safe operation of the heaters
- Prepared to install a printer
- Different types and combinations of options possible
- RS232-connection for PC Validation port
Benchtop electronic laboratory autoclaves (EL)
With higher capacity and better performance, Tuttnauer's EL Laboratory line of fully automatic, microprocessor controlled, benchtop steam sterilisers provides safe, economical and effective sterilisation. Integrated reservoir for water feed, with water level control and steam recirculation saves on demineralised water consumption and enables the autoclave to be independent of a water supply. External heating elements make the steriliser easy to clean.
Sterilization applications: liquids such as nutrient media and buffer solutions, solid items such as pipettes, tubes and filters, glassware and plastic articles and waste sterilisation.
Integrated water reservoir for demineralised water (requires manual filling)
Condensate is returned to integrated water reservoir
Easy loading and unloading
Heating elements around the chamber; easy cleaning of chamber
Besides electricity, no other utilities needed on the standard version
Vertical electronic laboratory autoclaves (ELV)
The Tuttnauer vertical, top loading laboratory autoclave is available in chamber sizes from 23 to 160 liters. With higher capacity and better performance, Tuttnauer's ELV Laboratory line of fully automatic vertical steam sterilizers provides safe, economical and effective sterilization. The ELV models have an advanced microprocessor based control panel.
Sterilization applications: liquids such as nutrient media and buffer solutions, solid items such as pipettes, tubes and filters, glassware and plastic articles and biohazard sterilization
Filling of demineralised water by main supply
Optimal use of chamber volume
Heating element in bottom part of chamber
Drain necessary (optional collection vessel)
Swivel wheels with brakes (optional)"
Safety features: A safety device prevents the operator from opening the door when chamber is pressurised; steam is not allowed to enter the chamber when the door is open; a cycle cannot start if the door is open or not properly locked; the door will not open until the temperature is below a specified safe level.
Double independent monitoring: The combined electronic and mechanical monitoring guarantees accurate results. The operator has two independent means to monitor temperature and pressure.
Standards: Tuttnauer’s high quality laboratory autoclaves comply with the strictest international directives and standards:
Pressure Equipment: PED 97/23 EEC, EN 10028-7, ASME Code Sec VIII
Safety: IEC/UL/EN61010-1, IEC 61010-2-040, EN 61326
Sterilization: DIN 58951 Series, ISO 17665-1:2006
Quality System: ISO 9001:2000, ISO 13485:2003