β-(N-Morpholino)ethansulfonsäure (MES) Monohydrat freie Säure ≥98%, gereinigt, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
4812-02EA
1426.39
EUR
4812-02
4812-04
4812-06
4812-08
β-(N-Morpholino)ethansulfonsäure (MES) Monohydrat freie Säure ≥98%, gereinigt, J.T.Baker®
MES (β-(N-Morpholino)ethansulfonsäure) Monohydrat
Suitable for use in biopharmaceutical manufacturing applications.
Formel:
C₆H₁₃NO₄S·1H₂O Schmelzpunkt: 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149409 CAS-Nummer: 145224-94-8 EINECS: 224-632-3 Merck Index: 13,05929 |
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use | ≥98% |
Assay (C₆H₁₃NO₄S·H₂O) | ≥98 % |
Appearance | Passes Test |
Identification (by IR) | Passes Test |
RNase Activity | Passes Test |
DNase Activity (None Detected) | Passes Test |
Protease Activity | Passes Test |
Color of a 1M Alkaline Solution (Colorless) | Passes Test |
Loss on Drying at 130°C | 7 - 10 % |
Heavy Metals (as Pb) | ≤2 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...