β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)), Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
NAD+, Nicotinamid-Adenin-Dinucleotid, oxidiert, Nadid, Nicotinsäureamid-Adenin-Dinucleotid, β-Nicotinamid-Adenin-Dinucleotid
481911-10
481911-5
481911-10EA
631
EUR
481911-10
481911-5
β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)), Millipore®
β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)
Formel:
C₂₁H₂₇N₇O₁₄P₂ Molecular Weight: 663.43 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00036253 CAS-Nummer: 53-84-9 EINECS: 200-184-4 Merck Index: 13,06370 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...