β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)), Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: NAD+, Nicotinamid-Adenin-Dinucleotid, oxidiert, Nadid, Nicotinsäureamid-Adenin-Dinucleotid, β-Nicotinamid-Adenin-Dinucleotid

481911-10 481911-5
481911-10EA 631 EUR
481911-10 481911-5
β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)), Millipore®
β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)
Formel: C₂₁H₂₇N₇O₁₄P₂
Molecular Weight: 663.43 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00036253
CAS-Nummer: 53-84-9
EINECS: 200-184-4
Merck Index: 13,06370

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR