Benzo[b]fluoranthen

Lieferant: Merck

48490
48490.EA 150 EUR
48490.
Benzo[b]fluoranthen
Benzo[b]fluoranthen
Formel: C₂₀H₁₂
MDL: MFCD00010582
CAS-Nummer: 205-99-2
EINECS: 205-911-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f5bb55e77c62af099b68470c190955881c92e2c&vid=cb452750aaa124dfce506ee8090c79f55de4c4c5&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR