Fluoranthen

Lieferant: Merck
Achtung

48535
48535.EA 115 EUR
48535.
Fluoranthen
Fluoranthene
Formel: C₁₆H₁₀
Molecular Weight: 202,26 g/mol
MDL: MFCD00001184
CAS-Nummer: 206-44-0
EINECS: 205-912-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7c2fbbecd7f20b36235685c724b6cc6d350dcb5&vid=a7c2fbbecd7f20b36235685c724b6cc6d350dcb5&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR