Cyclohexanon ≥99,8% (durch GC), AR® ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

4872-08EA 249.7 EUR
4872-08
Cyclohexanon ≥99,8% (durch GC), AR® ACS, Macron Fine Chemicals™
Cyclohexanone
Formel: C₆H₁₀(=O)
Siedepunkt: 155,6 °C (1013 hPa)
Schmelzpunkt: –31 °C
Dichte: 0,9478 g/cm³ (20 °C)
Flammpunkt: 43 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001625
CAS-Nummer: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3,III
REACH: 01-2119453616-35
Merck Index: 13,02755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8701dac7edb70a452483cd4f1bd716ca29ba9aa&vid=0360daffe7606174162954ba58a6ba2b78e28dfb&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements ≥99.8%
For Laboratory,Research,or Manufacturing Use ≤10
ACS - Assay (C₆H₁₀O) (by GC, corrected for water) ≥99.8 %
Appearance (clear, colorless liquid) Passes Test
ACS - Color (APHA) ≤10
ACS - Acidity (as CH₃COOH) ≤0.010 %
ACS - Residue after Evaporation ≤0.05 %
ACS - Specific Gravity at 25°/25°C 0.940 - 0.950
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR