(4-Nitrophenyl)-β-D-glucopyranosid, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem

Synonyme: (4-Nitrophenyl)-β-D-glucosid

487507-1
487507-1EA 69.6 EUR
487507-1
(4-Nitrophenyl)-β-D-glucopyranosid, Sigma-Aldrich®
(4-Nitrophenyl)-β-D-glucopyranosid
Formel: C₁₂H₁₅NO₈
Molecular Weight: 301.25 g/mol
MDL: MFCD00006593
CAS-Nummer: 2492-87-7
EINECS: 219-661-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR