S-Nitrosoglutathione (GSNO) 95 HPLC_ASSAY_METHOD, Calbiochem®, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem

Synonyme: (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-(nitrosothio)-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, GSNO, Nitrosoglutathione, S-Nitroso-L-Glutathione

Inert_Gas_filled

487920-10 487920-50 487920-1
487920-10EA 114 EUR
487920-10 487920-50 487920-1
S-Nitrosoglutathione (GSNO) 95 HPLC_ASSAY_METHOD, Calbiochem®, Sigma-Aldrich®
S-Nitrosoglutathione (GSNO)

A utility carrier of nitric oxide.


Breaks down at physiological pH to produce nitric oxide. Causes smooth muscle relaxation and inhibits platelet aggregation. Induces apoptosis in mouse thymocytes.

Formel: C₁₀H₁₆N₄O₇S
Molecular Weight: 336.319 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00214341
CAS-Nummer: 57564-91-7

Bestellen

Spezifikation Testergebnisse

Purity (HPLC) ≥95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR