Calciumchlorid, wasserfrei 93,0-100,5%, CALTAC FCC, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Achtung

4880-32EA 0 EUR
4880-32
Calciumchlorid, wasserfrei 93,0-100,5%, CALTAC FCC, Macron Fine Chemicals™
Calciumchlorid

  • Food GMP manufactured product

Formel: CaCl₂
Siedepunkt: 1600 °C (1013 hPa)
Schmelzpunkt: 772 °C
Dichte: 2,15 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010903
CAS-Nummer: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Bestellen

Spezifikation Testergebnisse

Food GMP Manufactured Product 93.0 - 100.5%
Meets F.C.C. Requirements Passes Test
Preserve in Tight Containers Passes Test
CAUTION: For Manufacturing, processing or repackaging ≤0.02%
Bulk Food Chemical Passes Test
FCC - Assay (CaCl₂) 93.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
Solubility Passes Test
Acid-Insoluble Substances - Retained Mass ≤0.02 %
Acid-Insoluble Substances - No Particles >2mm Passes Test
Arsenic (As) ≤3 mg/kg
Fluoride (F) ≤0.004 %
FCC - Lead (Pb) ≤5 mg/kg
Magnesium and Alkali Salts ≤5.0 %
pH (1 in 20) 4.5 - 11.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR