Sterilisation indicators
Lieferant: PHAGOGENE
60150 60151
489-0013EA 161 EUR
489-0013 489-0014
Sterilisation indicators
Sterilisationsindikatoren
The indicator turns from yellow to blue upon autoclaving.
  • Non-sequential indicator
  • Non-toxic organic ink
  • Declination according to cycles of autoclaving: 'short cycle', 'standard cycle' or 'prion cycle'
Standards: Compliant with ISO standards.