Bowie-Dick type test pack
Lieferant: PHAGOGENE
60149
489-0039EA 28.9 EUR
489-0039
Bowie-Dick type test pack
Sterilisationsindikatoren
Bowie-Dick is ready-to-use test package for daily penetration test of steam for sterilisers with porous charge.
  • Yellow or blue free turn indicating the good extraction of the air
  • Non-toxic organic ink
Standards: Conforms to ISO 11140-4; BSI Kitemark certified.