Sterilisationsindikatorband
Lieferant: SÜDPACK MEDICA
7600012
489-0046EA 48.2 EUR
489-0046
Sterilisationsindikatorband
Sterilisationsindikatoren
Tapes indicators by dry heat sterilisation.
The tape (ethylene oxide) changes from white to black after 3,5 minutes at 134 °C.
Dry heat steriliser tape (poupinel) changes from green to different shades of brown after a period at a temperature between 155 and 180 °C.