Kupfer(II)bromid 50% in wässriger Lösung, AR®, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

4895-22EA 1662.76 EUR
4895-22
Kupfer(II)bromid 50% in wässriger Lösung, AR®, Macron Fine Chemicals™
Kupfer(II)bromid
Formel: Br₂Cu
Dichte: 1,50 kg (1 L)
MDL: MFCD00010970
CAS-Nummer: 7789-45-9
UN: 3264
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 49 - 51%
Appearance (dark brown/black solution) Passes Test
Assay (CuBr₂) 49 - 51 %
Insoluble Matter ≤0.005 %
Sulfate (SO₄) ≤0.005 %
Chloride (Cl) ≤0.15 %
Substances not Precipitated by H₂S (as SO₄) ≤0.10 %
Iron (Fe) ≤0.003 %
pH of 10% solution 2.7 - 3.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR