Supelco® Organic Reference Standard, Epicatechin gallate solution
Lieferant: Merck
49060-U
49060-UEA 113 EUR
49060-U
Supelco® Organic Reference Standard, Epicatechin gallate solution
Standards Organische Standards
  • GHS signal: GHS02, GHS06, GHS08
Standards: Epicatechin gallate solution is a Certified Reference Material (CRM) produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Bitte beachten P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311