Essigsäure ≥99,8% (durch GC) Eluentzusatz, für die LC-MS, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

49199-50ML
49199-50MLEA 71.26 EUR
49199-50ML
Essigsäure ≥99,8% (durch GC) Eluentzusatz, für die LC-MS, Fluka™
Essigsäure
Formel: H₃CCOOH
Siedepunkt: 118 °C (1,013 hPa)
Schmelzpunkt: 16 °C
Dichte: 1,05 g/cm³ (20 °C)
Flammpunkt: 40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00036152
CAS-Nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Vapor Density 2.07 (vs air)
Assay 99.5 - 100.5 %
Purity (GC) Min. 99.8 %
Solidification Point 16.3 - 16.7 °C
Water (Karl Fischer) Max. 0.2 %
Silver (Ag) Max. 0.01 ppm
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Barium (Ba) Max. 0.01 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.2 ppm
Cadmium (Cd) Max. 0.02 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.05 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.2 ppm
Germanium (Ge) Max. 0.05 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.01 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.02 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.05 ppm
Lead (Pb) Max. 0.02 ppm
Strontium (Sr) Max. 0.01 ppm
Titanium (Ti) Max. 0.01 ppm
Thallium (Tl) Max. 0.05 ppm
Vanadium (V) Max. 0.01 ppm
Zinc (Zn) Max. 0.05 ppm
Zirconium (Zr) Max. 0.1 ppm
Chloride (Cl) Max. 0.5 ppm
Phosphate (PO₄) Max. 0.5 ppm
Sulfate (SO₄) Max. 0.5 ppm
Suitability for the LC-MS complying
Acetic aNHydride (GC) Max. 0.01 %
APHA max. 10
Explosion Limit 4 %, 59 °F
Refractive Index n20/D 1.371 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR