EDTA Dinatriumsalz Dihydrat 99,0-101,0%, AR® ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

4931-20EA 0 EUR
4931-20
EDTA Dinatriumsalz Dihydrat 99,0-101,0%, AR® ACS, Macron Fine Chemicals™
EDTA Dinatriumsalz Dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Schmelzpunkt: 255 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00150037
CAS-Nummer: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 99.0 - 101.0%
ACS - Assay (C₁₀H₁₄N₂Na₂O₈·2H₂O) 99.0 - 101.0 %
Identification A Passes Test
Identification B Passes Test
Identification C Passes Test
ACS - Heavy Metals (as Pb) ≤0.005 %
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤0.01 %
ACS - Nitrilotriacetic Acid (by polarography) ≤0.1 %
ACS - pH of 5% Solution at 25°C 4.0 - 6.0
Certificate Provided Reports Actual Lot Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR