L(+)-Lysin Monohydrochlorid 98,5-101,5% (Trockenmasse), GenAR® USP, Multikompendial, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: (S)-(+)-Lysin Monohydrochlorid, H-Lys-OH.HCl, (S)-(+)-2,6-Diaminohexansäure Hydrochlorid, (S)-(+)-2,6-Diaminocapronsäure Hydrochlorid, L(+)-Lysin Hydrochlorid, (S)-(+)-2,6-Diaminohexansäure Monohydrochlorid, (S)-(+)-2,6-Diaminocapronsäure Monohydrochlorid, (S)-(+)-Lysin Hydrochlorid

4933-09EA 10038.94 EUR
4933-09 MACR4933-06
L(+)-Lysin Monohydrochlorid 98,5-101,5% (Trockenmasse), GenAR® USP, Multikompendial, Macron Fine Chemicals™
L(+)-Lysin Monohydrochlorid
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
Molecular Weight: 182.65 g/mol
MDL: MFCD00064564
CAS-Nummer: 657-27-2

Bestellen

Spezifikation Testergebnisse

Meets J.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
GMP Manufactured Product
Meets B.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₆H₁₄N₂O₂·HCl) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation [Å]²⁵ ^D (+) 20.4 - 21.4 Degree
USP - Loss on Drying at 105°C ≤0.4 %
USP - Residue on Ignition ≤0.1 %
USP - Sulfate (SO₄) ≤0.03 %
USP - Iron (Fe) ≤0.003 %
USP - Related Compounds - Individual Impurities ≤0.5 %
USP - Related Compounds - Total Impurities ≤2.0 %
USP - Chloride (Cl) 19.0 - 19.6 %
EP/BP - Assay (C₆H₁₄N₂O₂·HCl) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification E Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation [Å]²º^D (+) 21.0 - 22.5 Degree
EP/BP-Ninhydrin-Positive Substances-Each ≤0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤1.0 %
EP/BP - Sulfate (SO₄) ≤300 ppm
EP/BP - Ammonium (NH₄) ≤0.02 %
EP/BP - Iron (Fe) ≤30 ppm
EP/BP - Loss on Drying at 105°C ≤0.5 %
EP/BP - Ash (sulfated) ≤0.1 %
JP - Assay (C₆H₁₄N₂O₂·HCl) (dried basis) 98.5 - 101.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Optical Rotation (+) 19.0 - 21.5 Degree
JP - pH (1 in 10) 5.0 - 6.0
JP - Clarity and Color of Solution Passes Test
JP - Sulfate (SO₄) = 0.028 %
JP - Ammonium (NH₄) ≤0.02 %
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Arsenic (As) ≤2 ppm
JP - Related Substances Passes Test
JP - Loss on Drying at 105°C = 1.0 %
JP - Residue on Ignition = 0.10 %
Endotoxin Concentration, IU/mg, For Information Only 19.0 - 21.5 Degree
No Class 1,2,3 or other solvents are used or produced in the 5.0 - 6.0
manufacturing or purification of the product. Passes Test
Elemental Impurities (USP (232), EP 5.20) - Information on elemental = 0.02 %
impurities for this product is available on the associated Product = 10 ppm
Regulatory Data Sheet and elemental impurity profile report. = 2 ppm
Endotoxin Concentration, IU/mg For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR