L(+)-Valin 98,5-101,5 Dried USP, FCC, multi-compendial, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: (S)-(+)-Valin, (S)-(+)-2-Amino-3-methylbutansäure, (S)-(+)-2-Aminoisovaleriansäure, H-Val-OH

4936-06EA 0 EUR
4936-06 4936-09
L(+)-Valin 98,5-101,5 Dried USP, FCC, multi-compendial, Macron Fine Chemicals™
L(+)-Valin
Formel: C₅H₁₁NO₂
Molecular Weight: 117.15 g/mol
Siedepunkt: 217
Schmelzpunkt: 86
MDL: MFCD00064220
CAS-Nummer: 72-18-4
Merck Index: 13,09458

Bestellen

Spezifikation Testergebnisse

Meets B.P. Chemical Specifications
GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₅H₁₁NO₂) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation [Å]²⁵ ^D (+) 26.6 - 28.8 Degree
USP - pH 5.5 - 7.0
USP - Loss on Drying at 105°C ≤0.3 %
USP - Residue on Ignition ≤0.1 %
USP - Chloride (Cl) ≤0.05 %
USP - Sulfate (SO₄) ≤0.03 %
USP - Iron (Fe) ≤0.003 %
USP - Related Compounds - Individual Impurities ≤0.5 %
USP - Related Compounds - Total Impurities ≤2.0 %
EP/BP - Assay (C₅H₁₁NO₂) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation [Å]²º^D (+) 26.5 - 29.0 Degree
EP/BP-Ninhydrin-Positive Substances-Impurity B ≤0.4 %
EP/BP-Ninhydrin-Positive Substances-Each ≤0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤1.0 %
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (SO₄) ≤300 ppm
EP/BP - Ammonium (NH₄) ≤0.02 %
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Drying ≤0.5 %
EP/BP - Ash (sulfated) ≤0.1 %
JP - Assay (C₅H₁₁NO₂) (dried basis) 98.5 - 101.0 %
JP - Identification Passes Test
JP - Optical Rotation (+) 26.5 - 29.0 Degree
JP - pH 5.5 - 6.5
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) = 0.021 %
JP - Sulfate (SO₄) = 0.028 %
JP - Ammonium (NH₄) = 0.02 %
JP - Heavy Metals (as Pb) = 20 ppm
JP - Arsenic (As) = 2 ppm
JP - Related Substances Passes Test
JP - Loss on Drying at 105°C = 0.30 %
JP - Residue on Ignition = 0.10 %
Endotoxin Concentration, IU/mg, For Information Only = 0.5 %
Only Class 2 Solvents (Methanol) are likely to be present. All are = 0.1 %
below Option 1 limits. 98.5 - 101.0 %
Metallic Residues: No metal catalysts or metal reagents, as defined by Passes Test
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of 26.5 - 29.0 Degree
this material. 5.5 - 6.5
Endotoxin Concentration, IU/mg For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR