Octyl-β-D-thioglucopyranosid, Calbiochem®
Lieferant: Merck Millipore Calbiochem
Synonyme:
Octyl-thioglucoside, OTG, 1-S-Octyl-β-D-thioglucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol, N-Octyl-1-thio-Beta-D-glucopyranoside, Octyl β-D-thioglucopyranoside
494461-5
494461-5EA
564
EUR
494461-5
Octyl-β-D-thioglucopyranosid, Calbiochem®
Octyl-β-D-thioglucopyranosid
Formel:
C₁₄H₂₈O₅S Molecular Weight: 308,44 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00012189 CAS-Nummer: 85618-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...