Supelco® TraceCERT® Organic Reference Standard, ʟ-Cystine
Lieferant: Merck
49603-100MG
49603-100MGEA 86.4 EUR
49603-100MG
Supelco® TraceCERT® Organic Reference Standard, ʟ-Cystine
Standards Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff3b13afab1b8b80fab989349e648452a6f937df&vid=ff3b13afab1b8b80fab989349e648452a6f937df&prid=web&clid=SAPDAM