Glycerin, wasserfrei ≥99,5% (durch GC), Puriss. p.a. ACS, destilliert, Riedel-de Haën™

Lieferant: Honeywell Chemicals

Synonyme: 1,2,3-Propantriol, Glycerol

49770-2.5L 49770-1L 49770-250ML
49770-2.5LEA 375.77 EUR
49770-2.5L 49770-1L 49770-250ML
Glycerin, wasserfrei ≥99,5% (durch GC), Puriss. p.a. ACS, destilliert, Riedel-de Haën™
Glycerin

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications

Formel: HOCH₂CH(OH)CH₂OH
Siedepunkt: 290 °C (1,013 hPa)
Schmelzpunkt: 18 °C
Dichte: 1,26 g/cm³ (20 °C)
Flammpunkt: 160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004722
CAS-Nummer: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (GC) Min. 99.5 %
Density (D 20/4) 1.261 - 1.263
Refractive index (n 20/D) 1.473 - 1.475
Water (Karl Fischer) Max. 0.1 %
Residue on ignition Max. 0.005 %
Indifference complying
Calcium (Ca) Max. 0.0005 %
Cadmium (Cd) Max. 0.0001 %
Cobalt (Co) Max. 0.0001 %
Chromium (Cr) Max. 0.0001 %
Copper (Cu) Max. 0.0001 %
Iron (Fe) Max. 0.0001 %
Potassium (K) Max. 0.002 %
Magnesium (Mg) Max. 0.0001 %
Manganese (Mn) Max. 0.0001 %
Sodium (Na) Max. 0.002 %
Nickel (Ni) Max. 0.0001 %
Lead (Pb) Max. 0.0001 %
Zinc (Zn) Max. 0.0001 %
Heavy metals (as Pb) Max. 0.0002 %
Halogene compounds (as Cl) Max. 0.003 %
Ammonium (NH₄) Max. 0.0005 %
Chloride (Cl) Max. 0.0001 %
Sulfate (SO₄) Max. 0.001 %
Reaction against H₂SO₄ complying
Acrolein, Glucose + NH4-compounds complying
Fatty acids and ester (as butyric acid) Max. 0.05 %
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR