Organic reference standard, Neotame, analytical standard, Supelco®
Lieferant: Merck
49777-100MG
49777-100MGEA
83
EUR
49777-100MG
Organic reference standard, Neotame, analytical standard, Supelco®
Standards
Organische Standards
Neotame is a white colored, dipeptide methyl ester, which is found to be a highly potent and non-nutritive sweetener or flavour enhancer and hence finds applications in variety of foods. It is manufactured from aspartame and 3,3-dimethylbutyraldehyde in a one-step chemical synthesis process.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.