Organic reference standard, 4-Butylresorcinol, analytical standard, Supelco®
Lieferant: Merck
49797-100MG
49797-100MGEA 165 EUR
49797-100MG
Organic reference standard, 4-Butylresorcinol, analytical standard, Supelco®
Standards Organische Standards
4-n-butylresorcinol is a derivative of resorcinol and a potent human tyrosinase inhibitor. It may be used to decrease skin irritation and is also known to inhibit melanin production.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e31df5181832f654f9d0db12df0ad77bf3559a5b&vid=e31df5181832f654f9d0db12df0ad77bf3559a5b&prid=web&clid=SAPDAM