Petroleumbenzin, 35…60 °C, AR® ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

4980-26EA 0 EUR
4980-26
Petroleumbenzin, 35…60 °C, AR® ACS, Macron Fine Chemicals™
Petroleumbenzin
Siedepunkt: 35…60 °C (1013 hPa)
MDL: MFCD00081849
CAS-Nummer: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements Passes Test
For Laboratory,Research,or Manufacturing Use ≤10
ACS - Acidity Passes Test
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.001 %
Appearance and Color Passes Test
ACS - Boiling Range(Lower) 35 - 60 °C
ACS - Boiling Range(Upper) 35 - 60 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR