Organic reference standard, Pyrrole, analytical standard, Supelco®
Lieferant: Merck
49817-1ML
49817-5ML
49817-1MLEA
51.2
EUR
49817-1ML
49817-5ML
Organic reference standard, Pyrrole, analytical standard, Supelco®
Standards
Organische Standards
Pyrrole is used as one of the building blocks of biochemical macrocycles such as chlorins, porphyrins and corrins. It is also used for many technologically relevel applications such as in organometallic complexes and precursor molecules for self-assembled monolayers on metal surfaces.