Organic reference standard, Ammelide, analytical standard, Supelco®
Lieferant: Merck
49866-10MG
49866-10MGEA
83
EUR
49866-10MG
Organic reference standard, Ammelide, analytical standard, Supelco®
Standards
Organische Standards
Ammelide can be synthesised via hydrolysis of 2-amino-4,6-dichloro-s-triazine in the presence of acid or base or by the reaction between urea and cyanogen iodide at 130 to 140 degrees.
Verpackung: Bottomless glass bottle contents are inside inserted with fused cone.