Organic reference standard, Ammelide, analytical standard, Supelco®
Lieferant: Merck
49866-10MG
49866-10MGEA 83 EUR
49866-10MG
Organic reference standard, Ammelide, analytical standard, Supelco®
Standards Organische Standards
Ammelide can be synthesised via hydrolysis of 2-amino-4,6-dichloro-s-triazine in the presence of acid or base or by the reaction between urea and cyanogen iodide at 130 to 140 degrees.
Verpackung: Bottomless glass bottle contents are inside inserted with fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83a6365c0f00873f2cee56c24c710c768bfae6fe&vid=83a6365c0f00873f2cee56c24c710c768bfae6fe&prid=web&clid=SAPDAM