Riboflavin (Vitamin B2) E101 (i), SAFC®

Lieferant: Merck

Synonyme: 6,7-Dimethyl-9-D-ribitylisoalloxazin, Lactoflavin, 7,8-Dimethyl-10-(D-ribo-2,3,4,5-tetrahydroxypentyl)-benzo[g]pteridin-2,4(3H,10H)-dion, (-)-Riboflavin, Vitamin B2, Vitamin G

M-Clarity™ Qualitätsstufe = MQ300

5.00257.2500
5.00257.2500EA 0 EUR
5.00257.2500
Riboflavin (Vitamin B2) E101 (i), SAFC®
Riboflavin (Vitamin B2)
Formel: C₁₇H₂₀N₄O₆
Molecular Weight: 376,37 g/mol
Schmelzpunkt: 290 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005022
CAS-Nummer: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR