BAPTA tetracesium salt ≥95% (durch TLC) Ionen-Chelator, zur Kalziumbestimmung

Lieferant: Biotium
Achtung

50001
50001.EA 551 EUR
50001.
BAPTA tetracesium salt ≥95% (durch TLC) Ionen-Chelator, zur Kalziumbestimmung
BAPTA tetracesium salt

BAPTA and its derivatives are calcium chelators that are commonly used to form calcium buffers with well-defined calcium concentrations.


  • Kd (no Mg2 ): 0.59 uM, Kd (1 mM Mg2 ): 0.70 uM
  • White solid soluble in water
  • Store at 4°C


By injecting the chelators into cells or by incubating cells with the AM ester form of the chelators, one can control the cytosolic calcium concentration, an important means to study the roles of calcium. Key advantages of these calcium chelators include relative insensitivity toward intracellular pH change and fast release of calcium. Biotium offers several BAPTA chelators with calcium dissociation constants covering the biologically significant range from 10-7 to 10-2 M.

Formel: C₂₃H₂₄Cs₄N₂O₁₀
Molecular Weight: 1004 g/mol
CAS-Nummer: 480436-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR