5,5′-Dibromo BAPTA tetrapotassium salt ≥95% (durch TLC) Ionen-Chelator, zur Kalziumbestimmung

Lieferant: Biotium

Synonyme: 5,5′-Dibromo BAPTA 4k+, 5,5'-Dibromo-BAPTA

50004
50004.EA 284 EUR
50004.
5,5′-Dibromo BAPTA tetrapotassium salt ≥95% (durch TLC) Ionen-Chelator, zur Kalziumbestimmung
5,5′-Dibromo BAPTA tetrapotassium salt

BAPTA and its derivatives are calcium chelators that are commonly used to form calcium buffers with well-defined calcium concentrations. By injecting the chelators into cells or by incubating cells with the AM ester form of the chelators, one can control the cytosolic calcium concentration, an important means to study the roles of calcium.


  • Kd (no Mg2 ): 3.6 uM
  • Cell permeability: Membrane impermeant
  • White to off-white solid soluble in water
  • Store at 4°C


Key advantages of these calcium chelators include relative insensitivity toward intracellular pH change and fast release of calcium. Biotium offers several BAPTA chelators with calcium dissociation constants covering the biologically significant range from 10-7 to 10-2 M.

Formel: C₂₂H₁₈Br₂K₄N₂O₁₀
Molecular Weight: 786,59 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00036169
CAS-Nummer: 73630-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR