FLUO-3 pentaammonium salt ≥95% (durch TLC) fluoreszierender Indikator, zur Kalziumbestimmung

Lieferant: Biotium

50010
50010.EA 335 EUR
50010.
FLUO-3 pentaammonium salt ≥95% (durch TLC) fluoreszierender Indikator, zur Kalziumbestimmung
FLUO-3 pentaammonium salt

Fluo-3, pentaammonium is membrane-impermeant, but can be loaded into cells via microinjection or scrape loading.


  • Kd: 450 nM
  • λEx/λEm (low or high [Ca2+]) = 506/526nm
  • ε (506 nm) = 86,000 M-1cm-1
  • Cell permeability: Membrane impermeant
  • Orange red solid soluble in DMSO and water (pH >6)
  • Store at 4°C and protect from light, especially when in solution

Formel: C₃₆H₂₅Cl₂N₇O₁₃
Lagertemperatur: Kühlschrank
CAS-Nummer: 134907-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR