FLUO 3/AM ≥95% (durch HPLC) fluoreszierender Indikator, zur Kalziumbestimmung

Lieferant: Biotium

50013 50014 50016
50013.EA 429 EUR
50013. 50014. 50016.
FLUO 3/AM ≥95% (durch HPLC) fluoreszierender Indikator, zur Kalziumbestimmung
FLUO 3/AM

Membrane-permeant AM ester form of Fluo-3 that can be loaded into cells via incubation. Fluo-3 AM ester itself does not bind Ca2 , but it is readily hydrolyzed to Fluo-3 by endogenous esterases once the dye is inside the cells. An important application of Fluo-3 AM ester is its use in high throughput drug screening.


  • Abs/Em (after hydrolysis): 506/526 nm (low or high [Ca2 ] )
  • Extinction Coefficient (after hydrolysis): 86,000 M-1 cm-1 (506 nm)
  • Kd (Ca2 ) (after hydrolysis): 450 nM
  • Cell permeability: Membrane permeant
  • Orange red solid soluble in DMSO
  • Store -20°C and protect from light, especially when in solution


Because of the relatively low water solubility of the AM ester, Pluronic F-127, a mild detergent, is often used as a dispersing agent to facilitate dye loading. See the product information sheet for more information.

Formel: C₅₁H₅₀Cl₂N₂O₂₃
Lagertemperatur: Gefrierschrank
CAS-Nummer: 121714-22-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR