Rhod-2 trisodium salt ≥90% (durch HPLC) fluoreszierender Indikator, zur Kalziumbestimmung
Lieferant: Biotium
50022
50022.EA
335
EUR
50022.
Rhod-2 trisodium salt ≥90% (durch HPLC) fluoreszierender Indikator, zur Kalziumbestimmung
Rhod-2 trisodium salt
Membrane-impermeant calcium indicator that can be loaded into cells via microinjection or scrape loading.
- Kd: 1 uM
- λEx/λEm (low or high [Ca2+])= 556/576 nm
- ε (556 nm) = 80,000 M-1cm-1
- Cell permeability: Membrane impermeant
- Orange red solid soluble in DMSO and water (pH >6)
- Store at 4°C and protect from light, especially in solution