Rhod-2 AM (Bromide) fluoreszierender Indikator, zur Kalziumbestimmung

Lieferant: Biotium

50024 50023
50024.EA 369 EUR
50024. 50023.
Rhod-2 AM (Bromide) fluoreszierender Indikator, zur Kalziumbestimmung
Rhod-2 AM (Bromide)

Membrane-permeant form of Rhod-2 that can be loaded into cells via incubation. Because of the relatively low water solubility of the AM ester, the mild detergent Pluronic F-127 is often used as a dispersing agent to facilitate dye loading. Rhod-2-AM itself does not bind Ca2+ , but it is readily hydrolyzed to Rhod-2 by endogenous esterases once it enters cells.


  • Absorption/Emission 556nm/576nm (low or high [Ca2+] )
  • Extinction Coefficient 80,000 M-1 cm-1 (556 nm)
  • Cell permeability: Membrane permeant
  • Red solid soluble in DMSO
  • Store at -20°C and protect from light, especially in solution

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR