Rhod-590 tripotassium salt fluoreszierender Indikator, zur Kalziumbestimmung
Lieferant: Biotium
50026
50026.EA 509 EUR
50026.
Rhod-590 tripotassium salt fluoreszierender Indikator, zur Kalziumbestimmung
Rhod-590 tripotassium salt
Membrane-impermeant calcium indicator that can be loaded into cells via microinjection or scrape loading.
  • Kd: 0.61 uM
  • λEx/λEm 590/616 nm for zero calcium and 595/626 nm for high calcium
  • Cell permeability: Membrane impermeant
  • Purple solid soluble in DMSO and water (pH >6)
  • Store at 4°C and protect from light, especially in solution
For the membrane-permeable AM ester form of Rhod-590, see 50025.