Tungsten Standard for ICP, TraceCERT®, Supelco®
Lieferant: Merck
50334-100ML
50334-100MLEA
123
EUR
50334-100ML
Tungsten Standard for ICP, TraceCERT®, Supelco®
Standards
Kationischer Standards
Wolfram-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034